Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCFD1 Antibody (OTI4D2), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB274003A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274003A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274003A48 Supplier Novus Biologicals Supplier No. NBP274003AF488

Mouse Monoclonal Antibody

SCFD1 Monoclonal specifically detects SCFD1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SCFD1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4D2
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C14orf163, KIAA0917chromosome 14 open reading frame 163, sec1 family domain containing 1, sec1 family domain-containing protein 1, SLY1, SLY1 homolog, sly1p, STXBP1L2RA410, Syntaxin-binding protein 1-like 2, vesicle transport-related protein
Gene Symbols SCFD1
Host Species Mouse
Immunogen Full length recombinant protein of human SCFD1 (NP_057190) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23256.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.