Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCFD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179522
Description
SCFD2 Polyclonal specifically detects SCFD2 in Mouse samples. It is validated for Western Blot.Specifications
SCFD2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ21060, FLJ39514, sec1 family domain containing 2, STXBP1L1sec1 family domain-containing protein 2, syntaxin binding protein 1-like 1, Syntaxin-binding protein 1-like 1 | |
Rabbit | |
52 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Dog: 86%; Pig: 86%; Bovine: 86%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_848787 | |
Scfd2 | |
Synthetic peptide directed towards the middle region of human Scfd2The immunogen for this antibody is Scfd2. Peptide sequence LVSGLSSLCEHLGVREECFAVGPLSRVIATDLANYAPAKNRKKTATGRAS. | |
Affinity purified | |
RUO | |
152579 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction