Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCGB1D2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106048 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106048 Supplier Novus Biologicals Supplier No. NBP106048
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SCGB1D2 Polyclonal specifically detects SCGB1D2 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen SCGB1D2
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006542.1
Gene Alias LIPB, LIPHB, lipophilin-B, LPHB, prostatein-like lipophilin B, secretoglobin family 1D member 2, secretoglobin, family 1D, member 2
Gene Symbols SCGB1D2
Host Species Goat
Immunogen Peptide with sequence CTDQMSLQKRSLIAE corresponding to internal region according to NP_006542.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10647
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.