Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCGF/CLEC11a Antibody (MM0533-6D5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018931 0.025 mg
NBP211874 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018931 Supplier Novus Biologicals Supplier No. NBP2118740.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCGF/CLEC11a Monoclonal specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SCGF beta
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Monoclonal
Clone MM0533-6D5
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q9Y240
Gene Alias CLECSF3C-type lectin superfamily member 3, C-type lectin domain family 11 member A, C-type lectin domain family 11, member A, LSLCLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 3, Lymphocyte secreted C-type lectin, lymphocyte secreted long form of C-type lectin, p47, SCGF, Stem cell growth factor, stem cell growth factor; lymphocyte secreted C-type lectin
Gene Symbols CLEC11A
Host Species Mouse
Immunogen Human SCGF beta
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6320
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.