Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCGF/CLEC11a Antibody (MM0533-6D5) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen SCGF beta
Clone MM0533-6D5
Concentration LYOPH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018931
SDP
View Documents
Novus Biologicals
NBP2118740.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP211874
SDP
View Documents
Novus Biologicals
NBP211874
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

SCGF/CLEC11a Monoclonal specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SCGF beta
LYOPH
Monoclonal
Purified
RUO
Q9Y240
CLEC11A
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
MM0533-6D5
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Unconjugated
Mouse
Human
6320
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.