Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SCH 221510
GREENER_CHOICE

Catalog No. 324010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-401-0 10 mg
32-405-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-401-0 Supplier Tocris Bioscience™ Supplier No. 3240/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective NOP agonist

Potent and selective nociceptin opioid receptor (NOP) agonist (EC50 values are 12, 693, 683 and 8071 nM, and Ki values are 0.3, 65, 131 and 2854 nM at NOP, μ-, κ- and δ-opioid receptors respectively). Exhibits anxiolytic-like activity at doses that have no effect on overt behaviors such as locomotion in vivo. Orally active.

Chemical Identifiers

CAS 322473-89-2
Molecular Formula C28H31NO

Specifications

Quantity 10 mg
Formula Weight 397.55
Purity >99%
Molecular Weight (g/mol) 397.562
Chemical Name or Material SCH 221510
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.