Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SCH 39166 hydrobromide
GREENER_CHOICESDP

Catalog No. 229950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-995-0 50 mg
22-991-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-995-0 Supplier Tocris Bioscience™ Supplier No. 2299/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity D1-like antagonist

High affinity dopamine D1/D5 receptor antagonist; displays Ki values of 1.2, 2, 980, 5520, 80 and 731 nM for binding to D1, D5, D2, D4, 5-HT and α2a receptors, respectively.

Chemical Identifiers

CAS 1227675-51-5
Molecular Formula C19H21BrClNO

Specifications

Quantity 50 mg
Formula Weight 394.73
Purity >99%
Molecular Weight (g/mol) 394.737
Chemical Name or Material SCH 39166 hydrobromide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.