Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Schistosoma Mansoni Antibody (BGN/1A7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017612 0.025 mg
NB10064566 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017612 Supplier Novus Biologicals Supplier No. NB100645660.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Schistosoma Mansoni Monoclonal specifically detects Schistosoma Mansoni in Invertebrate samples. It is validated for ELISA, Immunohistochemistry (Frozen).

Specifications

Antigen Schistosoma Mansoni
Applications ELISA, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone BGN/1A7
Concentration 1.5 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation PBS (pH 7.2) with 0.1% Sodium Azide
Host Species Mouse
Immunogen Native S. mansoni cercariae.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with antigen present in the urine of patients with Schistosomiasis. Reacts with a TCA soluble adult worm antigen in IF/ELISA.
Target Species Invertebrate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.