Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Schistosoma Mansoni Antibody (BGN/1A7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Schistosoma Mansoni |
---|---|
Clone | BGN/1A7 |
Concentration | 1.5 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunocytochemistry, Immunofluorescence, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017612
![]() |
Novus Biologicals
NB100645660.025MG |
0.025 mg |
Each for $181.90
|
|
|||||
NB10064566
![]() |
Novus Biologicals
NB10064566 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Schistosoma Mansoni Monoclonal specifically detects Schistosoma Mansoni in Invertebrate samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Schistosoma Mansoni | |
1.5 mg/mL | |
Immunocytochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Mouse | |
Invertebrate | |
IgG1 | |
Protein A purified | |
Reacts with antigen present in the urine of patients with Schistosomiasis. Reacts with a TCA soluble adult worm antigen in IF/ELISA. |
BGN/1A7 | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Native S. mansoni cercariae. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title