Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Schistosoma Mansoni Antibody (BGN/1A7), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10064566G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064566G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064566G Supplier Novus Biologicals Supplier No. NB10064566G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Schistosoma Mansoni Monoclonal specifically detects Schistosoma Mansoni in Invertebrate samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Schistosoma Mansoni
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone BGN/1A7
Conjugate DyLight 488
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Gene Alias S. mansoni
Host Species Mouse
Immunogen Native S. mansoni cercariae.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with antigen present in the urine of patients with Schistosomiasis. Reacts with a TCA soluble adult worm antigen in IF/ELISA.
Target Species Invertebrate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.