Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Schlafen 11 Rabbit anti-Human, Clone: 5P3W6, Novus Biologicals™
SDP

Catalog No. NB167267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167267 100 μg
NB167268 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167267 Supplier Novus Biologicals Supplier No. NBP316086100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Schlafen 11 Monoclonal antibody specifically detects Schlafen 11 in Human samples. It is validated for Western Blot

Specifications

Antigen Schlafen 11
Applications Western Blot
Classification Monoclonal
Clone 5P3W6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FLJ34922, schlafen family member 11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 132-328 of human Schlafen 11 (NP_689483.3).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91607
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.