Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCIMP Antibody (NVL-07), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP222349A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP222349A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP222349A32 Supplier Novus Biologicals Supplier No. NBP222349AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCIMP Monoclonal specifically detects SCIMP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SCIMP
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone NVL-07
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias transmembrane protein C17orf87, C17orf87, chromosome 17 open reading frame 87, DTFT5783, FLJ32580, SLP adapter and CSK-interacting membrane protein, SLP adaptor and CSK interacting membrane protein, SLP65/SLP76, Csk-interacting membrane protein, UNQ5783
Gene Symbols SCIMP
Host Species Mouse
Immunogen Recombinant intracellular part of human SCIMP.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 388325
Test Specificity The mouse monoclonal antibody NVL-07 recognizes intracellular part of human transmembrane adaptor SCIMP. This protein of 17 kDa predicted Mw migrates as a 22 kDa band on SDS PAGE.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.