Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCIMP Antibody (NVL-07), FITC, Novus Biologicals™
SDP

Catalog No. NB355705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355705 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355705 Supplier Novus Biologicals Supplier No. NBP222349F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCIMP Monoclonal antibody specifically detects SCIMP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Flow (Intracellular), CyTOF

Specifications

Antigen SCIMP
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone NVL-07
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Flow (Intracellular), CyTOF-ready
Formulation PBS
Gene Alias transmembrane protein C17orf87, C17orf87, chromosome 17 open reading frame 87, DTFT5783, FLJ32580, SLP adapter and CSK-interacting membrane protein, SLP adaptor and CSK interacting membrane protein, SLP65/SLP76, Csk-interacting membrane protein, UNQ5783
Host Species Mouse
Immunogen Recombinant intracellular part of human SCIMP.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388325
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.