Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCL/Tal1 Antibody (1E6), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250285UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250285UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250285UV Supplier Novus Biologicals Supplier No. NBP250285UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCL/Tal1 Monoclonal specifically detects SCL/Tal1 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SCL/Tal1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1000000
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product
Gene Symbols TAL1
Host Species Mouse
Immunogen GST-SCL/Tal1 (N-terminal) fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6886
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.