Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCL/Tal1 Antibody (BTL73), Biotin, Novus Biologicals™
SDP

Catalog No. NB385546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385546 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385546 Supplier Novus Biologicals Supplier No. NBP308616B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen SCL/Tal1
Applications Western Blot, ELISA
Classification Monoclonal
Clone BTL73
Conjugate Biotin
Formulation PBS
Gene Alias BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product
Host Species Mouse
Immunogen Recombinant human full-length TAL1 protein (Uniprot: P17542)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6886
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.