Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCMH1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220269 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220269 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220269 Supplier Novus Biologicals Supplier No. NBP220269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCMH1 Polyclonal specifically detects SCMH1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen SCMH1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:20000, Immunoprecipitation 1:10-1:500
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96GD3
Gene Alias polycomb protein SCMH1, Scml3, Sex comb on midleg homolog 1, sex comb on midleg homolog 1 (Drosophila)
Gene Symbols SCMH1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SCMH1. The exact sequence is proprietary.
Molecular Weight of Antigen 73 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22955
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.