Learn More
Description
Specifications
Specifications
| Antigen | Scn2b |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | neuronal voltage-gated sodium channel beta 2 subunit, sodium channel beta 2 subunit, sodium channel subunit beta-2, sodium channel, voltage-gated, type II, beta, sodium channel, voltage-gated, type II, beta polypeptide |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Scn2b (O60939; Met1-Ala159). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
