Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scn2b Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB329373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329373 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329373 Supplier Novus Biologicals Supplier No. NBP300206APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Scn2b Polyclonal antibody specifically detects Scn2b in Human samples. It is validated for ELISA

Specifications

Antigen Scn2b
Applications ELISA
Classification Polyclonal
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias neuronal voltage-gated sodium channel beta 2 subunit, sodium channel beta 2 subunit, sodium channel subunit beta-2, sodium channel, voltage-gated, type II, beta, sodium channel, voltage-gated, type II, beta polypeptide
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Scn2b (Accession#: O60939; Met1-Ala159)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6327
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.