Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCN3B Antibody (S396-29), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB259318488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259318488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259318488 Supplier Novus Biologicals Supplier No. NBP259318AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCN3B Monoclonal specifically detects SCN3B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SCN3B
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S396-29
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HSA243396, KIAA1158, SCNB3, sodium channel subunit beta-3, sodium channel, voltage-gated, type III, beta, voltage-gated sodium channel beta-3 subunit
Gene Symbols SCN3B
Host Species Mouse
Immunogen Fusion protein amino acids 1-215 (full-length) of rat NavBeta3
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 55800
Test Specificity Detects 40kDa. Does not cross-reat with NavBeta1, Navbeta2, or Navbeta4.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.