Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCN3B Antibody (S396-29), FITC, Novus Biologicals™
SDP

Catalog No. NBP259318F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259318F 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259318F Supplier Novus Biologicals Supplier No. NBP259318F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCN3B Monoclonal specifically detects SCN3B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SCN3B
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S396-29
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias HSA243396, KIAA1158, SCNB3, sodium channel subunit beta-3, sodium channel, voltage-gated, type III, beta, voltage-gated sodium channel beta-3 subunit
Gene Symbols SCN3B
Host Species Mouse
Immunogen Fusion protein amino acids 1-215 (full-length) of rat NavBeta3
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 55800
Test Specificity Detects 40kDa. Does not cross-reat with NavBeta1, Navbeta2, or Navbeta4.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.