Learn More
KIMBLE™ Score-Break Ampule, Trimmed Stem, Lab Pack
SDP
Score-break ampule with pre-scored constricted portion of the stem permits safe and easy opening while eliminating the need for filing.
$562.00 - $1302.00
Specifications
Material Borosilicate Glass (Type I)
Product Type Prescored Ampule
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
01-672-724
SDP
View Documents
KIMBLE™
12010L1
2 mL
Case of 1152 for $562.00
Only null left
Add to Cart
01-672-725
SDP
View Documents
KIMBLE™
12040U20
1 mL
Case of 1152 for $1,147.00
Only null left
Add to Cart
01-672-726
SDP
View Documents
KIMBLE™
12040U5
20 mL
Case of 3456 for $1,302.00
Only null left
Add to Cart
Description
Description
  • Converted from Type I borosilicate glass tubing, ampules conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1.
  • Pre-scored constricted portion of the stem permits safe and easy opening while eliminating the need for filing
  • Amber glass is recommended for light-sensitive compound
  • Stems can be pull or tip-sealed
  • Excellent dimensional stability for high speed filling and sealing operations
  • Customization including barcoding, pre-cleaning, and other capacities and styles is available; contact Customer Service
Specifications
Specifications
Borosilicate Glass (Type I)
Prescored Ampule
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.