Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300228 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300228 0.1 mL
NB300228SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300228 Supplier Novus Biologicals Supplier No. NB300228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCP1 Polyclonal specifically detects SCP1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.

Specifications

Antigen SCP1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot reported in scientific literature (PMID 23843895), Simple Western 1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:300-1:750, Immunohistochemistry-Paraffin reported in scientific literature (PMID 26858254), Immunoblotting reported in scientific literature (PMID 27431324)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
Gene Accession No. Q15431
Gene Alias Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1
Gene Symbols SYCP1
Host Species Rabbit
Immunogen A C-terminal synthetic peptide made to the human SCP1 protein sequence. [UniProt# Q15431]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6847
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.