Learn More
Description
Specifications
Specifications
| Antigen | SCP1 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot reported in scientific literature (PMID 23843895), Simple Western 1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:300-1:750, Immunohistochemistry-Paraffin reported in scientific literature (PMID 26858254), Immunoblotting reported in scientific literature (PMID 27431324) |
| Formulation | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide |
| Gene Accession No. | Q15431 |
| Gene Alias | Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 |
| Gene Symbols | SYCP1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
