Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB354869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354869 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354869 Supplier Novus Biologicals Supplier No. NB300228MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCP1 Polyclonal antibody specifically detects SCP1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot

Specifications

Antigen SCP1
Applications Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoblot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting
Formulation 50mM Sodium Borate
Gene Alias Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1
Host Species Rabbit
Immunogen A C-terminal synthetic peptide made to the human SCP1 protein sequence. [UniProt# Q15431]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6847
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.