Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCP3/SYCP3 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB300232V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300232V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300232V Supplier Novus Biologicals Supplier No. NB300232V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Feline, Parasite samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.

Specifications

Antigen SCP3/SYCP3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q8IZU3
Gene Alias MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3
Gene Symbols SYCP3
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of the human SCP3 protein. [UniProt# Q8IZU3]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 50511
Test Specificity Human, rat, pig, bovine and mouse. Other species have not been tested.
Target Species Mouse, Rat, Pig, Bovine, Chicken, Feline, Parasite
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.