Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCP3/SYCP3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB354620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354620 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354620 Supplier Novus Biologicals Supplier No. NBP278133MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCP3/SYCP3 Polyclonal antibody specifically detects SCP3/SYCP3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen SCP3/SYCP3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3
Host Species Rabbit
Immunogen Partial synthetic peptide made to an N-terminal region of the human SCP3 protein (between amino acids 10-80) [UniProt Q8IZU3]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 50511
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.