Learn More
Simport™ Scientific Screw Cap for Externally Threaded Sample Tubes (T501 and T502)
Shop All Simport Scientific ProductsDescription
- Made of polypropylene
- Autoclavable
- For externally threaded tubes
- Tubes sold separately
Specifications
Specifications
| Includes | Silicone washer |
| Product Type | Screw Cap |
| Material | Polypropylene |
| Color | Orange |
| No. per Pack | 1000 Pk. |
| Sterility | Non-sterile |
| For Use With (Equipment) | T501 and T502 Series Sample Tube |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.