Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scribble Antibody (7C6.D10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP229765 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229765 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229765 Supplier Novus Biologicals Supplier No. NBP229765
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Scribble Monoclonal specifically detects Scribble in Human samples. It is validated for Western Blot.

Specifications

Antigen Scribble
Applications Western Blot
Classification Monoclonal
Clone 7C6.D10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000
Gene Accession No. Q14160
Gene Alias protein scribble homolog, CRIB1, SCRB1, SCRIB1, scribbled homolog (Drosophila), Vartul
Gene Symbols SCRIB
Host Species Mouse
Immunogen GST-tagged fusion protein corresponding to the PDZ2 domain of Scribble.
Molecular Weight of Antigen 220 kDa
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23513
Test Specificity PDZ2 domain of Scribble.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.