Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCRN3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB191916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB191916 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB191916 Supplier Novus Biologicals Supplier No. NBP282003AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCRN3 Polyclonal antibody specifically detects SCRN3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen SCRN3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias FLJ23142, MGC149597, secernin 3, secernin-3
Host Species Rabbit
Immunogen SCRN3 antibody was raised against a 15 amino acid synthetic peptide near the center of human SCRN3. The immunogen is located within amino acids 230 - 280 of SCRN3. Amino Acid Squence: CEGYKLLNKHKGNIT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 79634
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.