Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Colored Screw Caps PROMO

Catalog No. 02-681-358S24 Shop All Fisherbrand Products
Click to view available options
Color:
Amber
Blue
Green
Natural
Orange
Red
Violet
Yellow

Colored screw caps for use with non-sterile, skirted and conical bottom tubes.

  • Frosted writing surface
  • Can be pierced for sample removal
TRUSTED_SUSTAINABILITY

Specifications

Includes O-ring
Product Type Screw Cap with O-ring
Autoclavable Autoclavable
Material Polypropylene
Color Natural
Sterility Non-sterile
For Use With (Equipment) Skirted and Conical Bottom Tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.