Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCUBE2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB356342 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356342 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356342 Supplier Novus Biologicals Supplier No. NBP177186MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCUBE2 Polyclonal antibody specifically detects SCUBE2 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence

Specifications

Antigen SCUBE2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Cegb1, Cegf1, CEGP1, FLJ16792, FLJ35234, MGC133057, Protein CEGP1, signal peptide, CUB and EGF-like domain-containing protein 2, signal peptide, CUB domain, EGF-like 2
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human SCUBE2. The immunogen is located within amino acids 480 - 530 of SCUBE2. Amino Acid Squence: CSPLRNKQQKSNDSAFGD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57758
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.