Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCX Antibody (PCRP-SCXA-1D2), FITC, Novus Biologicals™
SDP

Catalog No. NB187922 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187922 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187922 Supplier Novus Biologicals Supplier No. NBP314175F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SCX Monoclonal antibody specifically detects SCX in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen SCX
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-SCXA-1D2
Conjugate FITC
Formulation PBS with 0.05% Sodium Azide
Gene Alias BHLHA41, BHLHa48, Class A Basic Helix-Loop-Helix Protein 41, Class A Basic Helix-Loop-Helix Protein 48, Class II BHLH Protein Scleraxis, Scleraxis Basic Helix-Loop-Helix Transcription Factor, Scleraxis BHLH Transcription Factor, Scleraxis Homolog (Mouse), Scleraxis Homolog A, Scleraxis Homolog A (Mouse), Scleraxis Homolog B, Scleraxis Homolog B (Mouse), SCXA, SCXB
Host Species Mouse
Immunogen Recombinant full-length human SCX protein (Uniprot: Q7RTU7)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 642658
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.