Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCY1 like 3 Antibody (OTI1A5), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N274025J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274025J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274025J646 Supplier Novus Biologicals Supplier No. NBP274025JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCY1 like 3 Monoclonal specifically detects SCY1 like 3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SCY1 like 3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1A5
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ezrin-binding partner PACE-1 (PACE-1), Ezrin-binding protein PACE-1, PACE1PACE-1, protein-associating with the carboxyl-terminal domain of ezrin, RP1-97P20.2, SCY1-like 3 (S. cerevisiae), SCY1-like protein 3
Gene Symbols SCYL3
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human SCY1 like 3 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57147.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.