Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCYL1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB043703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043703 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043703 Supplier Novus Biologicals Supplier No. NBP297454R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCYL1 Polyclonal specifically detects SCYL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SCYL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Coated vesicle-associated kinase of 90 kDa, CVAK90, GKLPTelomerase transcriptional element-interacting factor, likely ortholog of mouse N-terminal kinase-like protein, MGC78454, NKTL, N-terminal kinase-like protein, NTKLN-terminal kinase-like, P105, SCY1-like 1 (S. cerevisiae), TAPKTeratoma-associated tyrosine kinase, TEIFSCY1-like protein 1, Telomerase regulation-associated protein, telomerase transcriptional elements-interacting factor, TRAPHT019
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SCYL1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 57410
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.