Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCYL1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $510.84
Specifications
Antigen | SCYL1 |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABS1494-25UG
|
MilliporeSigma
ABS149425UG |
25 μg |
Each for $192.35
|
|
|||||
ABS1494-100
|
MilliporeSigma
ABS1494100UG |
100 μg |
Each for $510.84
|
|
|||||
Description
Anti-SCYL1 is a rabbit polyclonal antibody that detects SCY1-Like Protein 1 and has been tested for use in Immunocytochemistry and Western Blotting.Specifications
SCYL1 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Human, Mouse | |
SCYL1,CVAK90,GKLP,NTKL,TAPK,TEIF,TRAP,HT01 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Rabbit | |
Signaling | |
NP_001041683.1 | |
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human SCY1-Like Protein 1. | |
Affinity Purified | |
This rabbit polyclonal antibody detects SCY1-Like Protein 1 (SCYL1) in human cells. It targets an epitope within 16 amino acids from the C-terminal region. The immunogen is conserved between isoforms 1, 2, and 3. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title