Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCYL1BP1 Antibody (OTI4F11), HRP, Novus Biologicals™
SDP

Catalog No. NBP274029H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274029H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274029H Supplier Novus Biologicals Supplier No. NBP274029H

Mouse Monoclonal Antibody

SCYL1BP1 Monoclonal specifically detects SCYL1BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SCYL1BP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4F11
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias FLJ11752, golgin, RAB6-interacting, GONTKL-binding protein 1, hNTKL-BP1, MGC51263, MGC70512, N-terminal kinase-like-binding protein 1, NTKL-BP1, NTKLBP1SCY1-like 1-binding protein 1, RAB6-interacting golgin, SCY1-like 1 binding protein 1, SCYL1-BP1, SCYL1BP1SCYL1-binding protein 1
Gene Symbols GORAB
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 35-394 of human SCYL1BP1 (NP_689494) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 92344.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.