Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCYL1BP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP260715 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260715 Supplier Novus Biologicals Supplier No. NBP260715

Rabbit Polyclonal Antibody

SCYL1BP1 Polyclonal specifically detects SCYL1BP1 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen SCYL1BP1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation 8-15 ul/mg lysate
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias FLJ11752, golgin, RAB6-interacting, GONTKL-binding protein 1, hNTKL-BP1, MGC51263, MGC70512, N-terminal kinase-like-binding protein 1, NTKL-BP1, NTKLBP1SCY1-like 1-binding protein 1, RAB6-interacting golgin, SCY1-like 1 binding protein 1, SCYL1-BP1, SCYL1BP1SCYL1-binding protein 1
Gene Symbols GORAB
Host Species Rabbit
Immunogen between 325 and 375
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 92344
Target Species Human
Content And Storage Store at 4degreesC.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.