Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SD 208
GREENER_CHOICE

Catalog No. 326910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-691-0 10 mg
32-695-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-691-0 Supplier Tocris Bioscience™ Supplier No. 3269/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent ATP-competitive TGF-βRI inhibitor

Potent, orally active ATP-competitive transforming growth factor-β receptor 1 (TGF-βRI) inhibitor (IC50 = 49 nM). Displays > 100-fold and > 17-fold selectivity over TGF-βRII and other common kinases respectively. Exhibits anti-inflammatory and antitumor activity. Also promotes an antiglioma immune response. Ameliorates germinal matrix hemorrhage-induced neurological deficits in neonatal rats.

Chemical Identifiers

CAS 627536-09-8
Molecular Formula C17H10ClFN6

Specifications

Quantity 10 mg
Formula Weight 352.75
Purity >98%
Molecular Weight (g/mol) 352.76
Chemical Name or Material SD 208
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.