Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDCCAG10 Antibody (OTI4C8), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274031FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274031FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274031FR Supplier Novus Biologicals Supplier No. NBP274031FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SDCCAG10 Monoclonal specifically detects SDCCAG10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SDCCAG10
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4C8
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Antigen NY-CO-10, CWC27 spliceosome-associated protein homolog (S. cerevisiae), EC 5.2.1.8, NY-CO-10, peptidyl-prolyl cis-trans isomerase SDCCAG10, PPIase CWC27, PPIase SDCCAG10, SDCCAG10, Serologically defined colon cancer antigen 10peptidyl-prolyl cis-trans isomerase CWC27 homolog
Gene Symbols CWC27
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 145-472 of human SDCCAG10 (NP_005860) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10283.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.