Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDF2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB088862 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088862 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088862 Supplier Novus Biologicals Supplier No. NBP299669AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SDF2 Polyclonal specifically detects SDF2 in Human samples. It is validated for ELISA.

Specifications

Antigen SDF2
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias SDF-2, stromal cell-derived factor 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SDF2 (NP_008854.2; Met1-Leu211).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6388
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.