Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDHA Rabbit anti-Human, Clone: JM10-83, Novus Biologicals™
SDP

Catalog No. NBP267130 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267130 Supplier Novus Biologicals Supplier No. NBP267130

Rabbit Monoclonal Antibody

SDHA Monoclonal specifically detects SDHA in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SDHA
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JM10-83
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:5000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias CMD1GG, EC 1.3.5.1, Fp, SDH1, SDH2, SDHFmitochondrial, succinate dehydrogenase complex flavoprotein subunit, succinate dehydrogenase complex, subunit A, flavoprotein (Fp)
Gene Symbols SDHA
Host Species Rabbit
Immunogen Synthetic peptide within Human SDHA aa 581-624/ 664. (SwissProt: P31040 Human; SwissProt: Q8K2B3 Mouse; SwissProt: Q920L2 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 6389
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.