Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDHB Antibody (SDHB/6697R), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB384214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384214 Supplier Novus Biologicals Supplier No. NBP308827IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SDHB Monoclonal antibody specifically detects SDHB in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SDHB
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SDHB/6697R
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in human SDHB was used as an immunogen. (Uniprot: P21912 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6390
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.