Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDHD Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341755 Supplier Novus Biologicals Supplier No. NBP281948MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SDHD Polyclonal antibody specifically detects SDHD in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SDHD
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CBT1, CII-4, CybS, PGL, PGL1, QPs3, SDH4, succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial, Succinate dehydrogenase complex subunit D, succinate dehydrogenase complex, subunit D, integral membrane protein, succinate dehydrogenase ubiquinone cytochrome B small subunit, Succinate-ubiquinone oxidoreductase cytochrome b small subunit, Succinate-ubiquinone reductase membrane anchor subunit
Host Species Rabbit
Immunogen SDHD antibody was raised against a 15 amino acid synthetic peptide near the center of human SDHD. The immunogen is located within amino acids 30 - 80 of SDHD. Amino Acid Squence: HLSPSHHSGSKAASL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6392
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.