Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDR16C5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP187150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP187150 0.1 mL
NB406199 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP187150 Supplier Novus Biologicals Supplier No. NBP187150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

SDR16C5 Polyclonal specifically detects SDR16C5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SDR16C5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias EC 1.1.1.105, EPHD-2, epidermal retinol dehydrogenase 2, FLJ33105, RDH-E2epidermal retinal dehydrogenase 2, RDHE2RDH#2, retinal short chain dehydrogenase reductase, Retinal short-chain dehydrogenase reductase 2, retSDR2, short chain dehydrogenase/reductase family 16C, member 5, Short-chain dehydrogenase/reductase family 16C member 5
Gene Symbols SDR16C5
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:CTTGCPSLLPILEPKYAVEKIVEAILQEKMYLYMPKLLYFMMFLKSFLPLKTGLLIADYLGILHAMDGFVDQKKKL
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 195814
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.