Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDR9C7 Antibody (OTI4B5), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274036FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274036FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274036FR Supplier Novus Biologicals Supplier No. NBP274036FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SDR9C7 Monoclonal specifically detects SDR9C7 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen SDR9C7
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI4B5
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 1.1.1, EC 1.1.1.-, FLJ16333, MGC126600, Orphan short-chain dehydrogenase/reductase, RDH-S, RDHSMGC126602, SDR-Oorphan short-chain dehydrogenase/ reductase, SDROretinol dehydrogenase similar protein, short chain dehydrogenase/reductase family 9C, member 7, short-chain dehydrogenase/reductase family 9C member 7
Gene Symbols SDR9C7
Host Species Mouse
Immunogen Full length recombinant protein of human SDR9C7 (NP_683695) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 121214.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.