Learn More
Bolle Safety™ Sealed Universal Safety Goggles
This goggle provides optimum protection and comfort in all circumstances thanks to its ergonomic design. This innovative goggle with its exclusive platinum coating is available in several versions, each adapted to a specific work environment to offer more peace of mind and efficiency for the worker.
$49.28 - $60.50
Specifications
Lens Coating Scratch-resistant, Anti-fog
Description Sealed
Product Type Safety Goggles
Material Polycarbonate
Disposable Reusable
View More Specs
Products 4
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
17-467-445
View Documents
Bolle Safety™
UNIVGN21W
Smoke and Foam
Each for $55.87
Only null left
Add to Cart
17-467-447
View Documents
Bolle Safety™
UNIVGN15W
Clear and Foam
Each for $60.50
Only null left
Add to Cart
17-467-449
View Documents
Bolle Safety™
UNIVGN13W
Clear with Neoprene Strap
Each for $49.28
Only null left
Add to Cart
17-467-450
View Documents
Bolle Safety™
UNIVGN11W
Clear
Each for $52.20
Only null left
Add to Cart
Description
Description
  • Optimal fit thanks to its Wave technology
  • PC/TPR co-injected frame with overflow chute
  • Effortless pivoting faceguard
  • Improved recyclability
  • UV protection
  • Scratch-resistant
  • Anti-fog
Specifications
Specifications
Scratch-resistant, Anti-fog
Safety Goggles
Reusable
Polycarbonate
Sealed
Polycarbonate
Polycarbonate
Standard
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.