Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Seamless Knit Nylon Gloves with Polyurethane Coated Fingertips

Catalog No. p-7219149 Shop All Showa Best Glove Products
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

General purpose gloves feature a snug fit and polyurethane coated fingertips for sensitivity and control. Permeable construction allows the hands to breathe and maintain temperature control.

  • Polyurethane fingertips provide touch sensitivity
  • Coating is permeable, allowing the hands to breathe
  • Deformation-resistant glove shape is maintained for an extended period of time
  • No fingerprints are left on clear, clean objects
  • Snug fit maximizes tactile feedback
  • Designed for precision applications

Specifications

Coated Area Fingertips
Construction Seamless Knit
Cuff Material Elastic
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
EN388 Standard 0131X
For Use With (Application) Electronic Components, Quality Control, and Integrated Circuits
Glove Cuff Color Natural
Glove Cuff Style Knit
Series BO600
Yarn Gauge 13
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.