Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seasonal H1N1 Hemagglutinin Antibody (7H12F6), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343350 Supplier Novus Biologicals Supplier No. NBP241328MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Seasonal H1N1 Hemagglutinin Monoclonal antibody specifically detects Seasonal H1N1 Hemagglutinin in Influenza virusN1, Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Seasonal H1N1 Hemagglutinin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7H12F6
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain
Host Species Mouse
Immunogen Antibody was raised against a synthetic peptide from the Hemagglutinin protein. Amino Acid Squence: PNIGDQKALTHTEN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus N1, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.