Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seasonal H1N1 Hemagglutinin Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB344125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344125 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344125 Supplier Novus Biologicals Supplier No. NBP241104MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Seasonal H1N1 Hemagglutinin Polyclonal antibody specifically detects Seasonal H1N1 Hemagglutinin in Influenza virusN1, Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Seasonal H1N1 Hemagglutinin
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide from the Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41106 and NBP2-41108. This antibody is a cognate pair with product NBP2-41105. The immunogen is located within amino acids 120 - 170 of Seasonal H1N1 Hemagglutinin. Amino Acid Squence: PNHTVTGVSAS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus N1, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.