Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seasonal H1N1 Neuraminidase Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396503 0.025 mg
NBP241111 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396503 Supplier Novus Biologicals Supplier No. NBP2411110.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Seasonal H1N1 Neuraminidase Polyclonal specifically detects Seasonal H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.

Specifications

Antigen Seasonal H1N1 Neuraminidase
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias EC=3.2.1.18, Neuraminidase
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) Neuraminidase protein. This antibody is a cognate pair with product NBP2-41112. The immunogen is located within the first 50 amino acids of Seasonal H1N1 Neuraminidase. Amino Acid Squence: VAGKDKTSVT
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for Neuraminidase from the seasonal influenza (H1N1) and will not recognize the corresponding Neuraminidase sequence from the swine-origin influenza virus (A/California/14/2009 (H1N1)). Will not cross-react with peptide corresponding to the swine-origin influenza (H1N1) Neuraminidase.
Target Species Influenza virus, Virus
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.